Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFI44L Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304507100UL
Description
IFI44L Polyclonal specifically detects IFI44L in Human, Mouse samples. It is validated for Western Blot.Specifications
IFI44L | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
10964 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
C1orf29, chromosome 1 open reading frame 29, GS3686, interferon-induced protein 44-like | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human IFI44L (NP_006811.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction