Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI44L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983874
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3874 Supplier Bioss Supplier No. BS15551R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function remains known.

Specifications

Antigen IFI44L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IFI44L
Gene Accession No. Q53G44
Gene Alias C1orf29; GS3686; H28; H-28; H28-1; H28c minor histocompatibility antigen 1; histocompatibility 28; HLA-HA28; IFI44L; IFL8; interferon induced protein 44 like; interferon-induced protein 44 like; Interferon-induced protein 44-like; Minor histocompatibility antigen HA-28; NS1178
Gene Symbols IFI44L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IFI44L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10964
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.