Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIH1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501733591 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3591 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3591 Supplier Proteintech Group Inc Supplier No. 217751AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Melanoma differentiation associated gene 5, MDA5, is a pattern recognition receptor to transmit the signal.Once activated, both proteins signal through IPS-1 activating transcription factors NF- kappaB and IRF-3 and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus, paromyxoviruses, Japanese encephalitis virus. MDA5 is essential for signaling by picornavirus. Both RIG-I and MDA5 are responsible for signaling by West Nile virus.

Specifications

Antigen IFIH1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFIH1
Gene Accession No. Q9BYX4
Gene Alias Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, IFIH1, MDA 5, MDA5, RH116, RLR-2
Gene Symbols IFIH1
Host Species Rabbit
Immunogen IFIH1 Fusion Protein Ag16467
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64135
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.