Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT1 Antibody (OTI3G8), Novus Biologicals™
SDP

Catalog No. NBP202340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202340 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202340 Supplier Novus Biologicals Supplier No. NBP202340
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFIT1 Monoclonal specifically detects IFIT1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen IFIT1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3G8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50-100, Immunohistochemistry-Paraffin 1:50
Gene Alias G10P1C56, GARG-16, IFI56IFI-56, IFI-56K, IFIT-1, IFNAI1ISG56, Interferon, alpha-inducible protein (MW 56kD), Interferon-induced 56 kDa protein, interferon-induced protein 56, interferon-induced protein with tetratricopeptide repeats 1, p56, RNM561
Gene Symbols IFIT1
Host Species Mouse
Immunogen Full length human recombinant protein of human IFIT1 (NP_000899) produced in HEK293T cell.
Molecular Weight of Antigen 55.2 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3434
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.