Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFIT1 Antibody (OTI3G8), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271005PCP
Description
IFIT1 Monoclonal specifically detects IFIT1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.Specifications
IFIT1 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
IFIT1 | |
Full length recombinant protein of human IFIT1 (NP_000899) produced in HEK293T cells. | |
0.1 mL | |
3434.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI3G8 | |
Flow Cytometry | |
G10P1C56, GARG-16, IFI56IFI-56, IFI-56K, IFIT-1, IFNAI1ISG56, Interferon, alpha-inducible protein (MW 56kD), Interferon-induced 56 kDa protein, interferon-induced protein 56, interferon-induced protein with tetratricopeptide repeats 1, p56, RNM561 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction