Learn More
IFIT1 Mouse anti-Human, Mouse, Rat, Clone: OTI3G8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500948
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Interferon-induced antiviral RNA-binding protein that specifically binds single-stranded RNA bearing a 5′-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs and inhibiting expression of viral messenger RNAs. Single-stranded PPP-RNAs, which lack 2′-O-methylation of the 5′ cap and bear a 5′-triphosphate group instead, are specific from viruses, providing a molecular signature to distinguish between self and non-self mRNAs by the host during viral infection. Directly binds PPP-RNA in a non-sequence-specific manner. Viruses evolved several ways to evade this restriction system such as encoding their own 2′-O-methylase for their mRNAs or by stealing host cap containing the 2′-O-methylation (cap snatching mechanism). Exhibits antiviral activity against several viruses including human papilloma and hepatitis C viruses.Specifications
| IFIT1 | |
| Monoclonal | |
| Unconjugated | |
| IFIT1 | |
| C56, G10P1, IFI-56, IFI-56K, IFI56, IFIT-1, IFNAI1, ISG56, P56, RNM561 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 15957, 3434, 56824 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P09914, Q64282 | |
| Ifit1 | |
| Full length human recombit protein of human IFIT1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.