Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT1 Mouse anti-Mouse, Unconjugated, Clone: 13B4-1, MilliporeSigma™

Catalog No. MABF22282MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22282MI 25 μg
MABF22281MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22282MI Supplier MilliporeSigma Supplier No. MABF222825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFIT1 Monoclonal antibody specifically detects IFIT1 in Mouse samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT1
Applications Western Blot
Classification Monoclonal
Clone 13B4-1
Conjugate Unconjugated
Gene Accession No. NP_032357
Gene Alias Interferon-induced protein with tetratricopeptide repeats 1;IFIT-1;Glucocorticoid-attenuated response gene 16 protein;GARG-16;Interferon-induced 56 kDa protein;IFI-56K;P56
Gene Symbols Ifit1, Garg16, Ifi56, Isg56
Host Species Mouse
Immunogen His-tagged full-length recombinant mouse Interferon-induced protein with tetratricopeptide repeats 1 (ITIT1).
Molecular Weight of Antigen ∼53 kDa observed; 53.74 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 13B4-1 specifically detects murine Interferon-induced protein with tetratricopeptide repeats 1 (IFIT1).
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.