Learn More
Invitrogen™ IFIT1 Polyclonal Antibody
Description
PA3-846 detects ISG56 (p56) from mouse samples. PA3-846 has been successfully used in Western blot applications to detect a 39-58 kDa band.
Specifications
Specifications
| Antigen | IFIT1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.05% sodium azide |
| Gene | IFIT1 |
| Gene Accession No. | Q64282 |
| Gene Alias | C56; G10P1; Garg16; GARG-16; glucocorticoid-attenuated response gene 16 product; glucocorticoid-attenuated response gene 16 protein; Ifi56; IFI-56; IFI-56K; IFIT1; IFIT-1; IFNAI1; interferon induced protein with tetratricopeptide repeats 1; interferon, alpha-inducible protein (MW 56kD); interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-induced protein with tetratricopeptide repeats 1-like protein; interferon-inducible mRNA 561; Isg56; P56; RNM561 |
| Gene Symbols | IFIT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.