Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531254 Supplier Invitrogen™ Supplier No. PA531254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HeLa(10 ng/mL hIFN-gamma1 treatment for 16hr). Predicted reactivity: Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Interferon-induced antiviral RNA-binding protein that specifically binds single-stranded RNA bearing a 5'-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs and inhibiting expression of viral messenger RNAs. Single-stranded PPP-RNAs, which lack 2'-O-methylation of the 5' cap and bear a 5'-triphosphate group instead, are specific from viruses, providing a molecular signature to distinguish between self and non-self mRNAs by the host during viral infection. Directly binds PPP-RNA in a non-sequence-specific manner. Viruses evolved several ways to evade this restriction system such as encoding their own 2'-O-methylase for their mRNAs or by stealing host cap containing the 2'-O-methylation (cap snatching mechanism). Exhibits antiviral activity against several viruses including human papilloma and hepatitis C viruses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene IFIT1
Gene Accession No. P09914
Gene Alias C56; G10P1; Garg16; GARG-16; glucocorticoid-attenuated response gene 16 product; glucocorticoid-attenuated response gene 16 protein; Ifi56; IFI-56; IFI-56K; IFIT1; IFIT-1; IFNAI1; interferon induced protein with tetratricopeptide repeats 1; interferon, alpha-inducible protein (MW 56kD); interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-induced protein with tetratricopeptide repeats 1-like protein; interferon-inducible mRNA 561; Isg56; P56; RNM561
Gene Symbols IFIT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IFIT1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.