Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA3848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3848 Supplier Thermo Scientific Supplier No. PA3848

Rabbit Polyclonal Antibody

PA3-848 detects ISG56 (p56) from human and mouse samples. PA3-848 has been successfully used in Western blot and ICC/IF applications to detect a 39-58 kDa band.

IFIT1 Polyclonal antibody specifically detects IFIT1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene IFIT1
Gene Accession No. P09914, Q64282
Gene Alias C56; G10P1; Garg16; GARG-16; glucocorticoid-attenuated response gene 16 product; glucocorticoid-attenuated response gene 16 protein; Ifi56; IFI-56; IFI-56K; IFIT1; IFIT-1; IFNAI1; interferon induced protein with tetratricopeptide repeats 1; interferon, alpha-inducible protein (MW 56kD); interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-induced protein with tetratricopeptide repeats 1-like protein; interferon-inducible mRNA 561; Isg56; P56; RNM561
Gene Symbols IFIT1
Host Species Rabbit
Immunogen Bacterially expressed full-length human p56 protein
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15957, 3434
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.