Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564079 Supplier Thermo Scientific Supplier No. PA564079
Add to Cart
Add to Cart

IFIT1B Polyclonal Antibody

IFIT1B Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFIT1B
Gene Accession No. Q5T764
Gene Alias 2010002M12Rik; AW412491; bA149I23.6; IFIT1B; IFIT1L; interferon induced protein with tetratricopeptide repeats 1B; Interferon-induced protein with tetratricopeptide repeats 1B; interferon-induced protein with tetratricopeptide repeats 1-like protein; RIKEN cDNA 2010002M12; RIKEN cDNA 2010002M12 gene
Gene Symbols IFIT1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFIT1B. Recombinant protein control fragment (Product #RP-103480).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 439996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.