Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFIT1B Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15554R

Description
IFIT1B gene ontology (GO) annotations include defense response to virus.Specifications
IFIT1B | |
Polyclonal | |
Unconjugated | |
IFIT1B | |
2010002M12Rik; AW412491; bA149I23.6; IFIT1B; IFIT1L; interferon induced protein with tetratricopeptide repeats 1B; Interferon-induced protein with tetratricopeptide repeats 1B; Interferon-induced protein with tetratricopeptide repeats 1-like protein; RIKEN cDNA 2010002M12; RIKEN cDNA 2010002M12 gene | |
Rabbit | |
Protein A | |
RUO | |
439996 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5T764 | |
IFIT1B | |
KLH conjugated synthetic peptide derived from human IFIT1B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction