Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFIT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528800
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IFIT2 belongs to the IFIT family. It contains 6 TPR repeats. IFIT genes comprise a large family with three (Ifit1, Ifit2, and Ifit3) and four (IFIT1, IFIT2, IFIT3, and IFIT5) members in mice and humans, respectively. IFITproteins are produced in human body that are supposed to confer immunity against viral infection. These proteins are generally produced during viral infection, Interferon (IFN) treatment, and during pathogen recognition (Pathogen associated molecular pattern recognition) byimmune systemduring infections.
Specifications
| IFIT2 | |
| Polyclonal | |
| Unconjugated | |
| Ifit2 | |
| AV302338; cig42; CIG-42; G10P2; Garg39; GARG-39; Glucocorticoid-attenuated response gene 39 protein; IFI54; IFI-54; IFI-54K; Ifit2; IFIT-2; interferon induced protein with tetratricopeptide repeats 2; Interferon, alpha-inducible protein (MW 54kD); interferon-induced 54 kDa protein; interferon-induced protein 54; interferon-induced protein with tetratricopeptide repeats 2; ISG54; ISG-54 K; ISG-54K; P54 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P09913 | |
| Ifit2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human IFIT2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction