Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFIT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589415
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589415 Supplier Invitrogen™ Supplier No. PA589415
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALEYIPNNAY LHCQIGCCYR AKVFQVMNLR ENGMYGKRKL LELIGHAVAH LKKADEANDN LFRVCSILAS LHALADQYED AEYYFQKEFS KELTPVAKQL LHLRYGNFQL YQMKCEDKAI HHFIEGVKIN QKSREKEKMK DKLQKIAKMR LSKNGADSEA LHVLAFLQEL NEKMQQADED SERGLESGSL IPSASSWNGE; Positive Samples: A-431, HepG2; Cellular Location: Cytoplasm, Endoplasmic reticulum.

IFIT2 belongs to the IFIT family. It contains 6 TPR repeats. IFIT genes comprise a large family with three (Ifit1, Ifit2, and Ifit3) and four (IFIT1, IFIT2, IFIT3, and IFIT5) members in mice and humans, respectively. IFITproteins are produced in human body that are supposed to confer immunity against viral infection. These proteins are generally produced during viral infection, Interferon (IFN) treatment, and during pathogen recognition (Pathogen associated molecular pattern recognition) byimmune systemduring infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ifit2
Gene Accession No. P09913
Gene Alias AV302338; cig42; CIG-42; G10P2; Garg39; GARG-39; Glucocorticoid-attenuated response gene 39 protein; IFI54; IFI-54; IFI-54K; Ifit2; IFIT-2; interferon induced protein with tetratricopeptide repeats 2; Interferon, alpha-inducible protein (MW 54kD); interferon-induced 54 kDa protein; interferon-induced protein 54; interferon-induced protein with tetratricopeptide repeats 2; ISG54; ISG-54 K; ISG-54K; P54
Gene Symbols Ifit2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 273-472 of human IFIT2 (NP_0015384).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3433
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.