Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFIT3 Antibody (OTI1G1), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271006JF646
Description
IFIT3 Monoclonal specifically detects IFIT3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IFIT3 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IFIT3 | |
Full length recombinant protein of human IFIT3 (NP_001540) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1G1 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49 | |
Mouse | |
Affinity Purified | |
Immunology | |
3437.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction