Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFIT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589571 Supplier Invitrogen™ Supplier No. PA589571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-29, COS-7, Mouse liver; Cellular Location: Cytoplasm, Mitochondrion Immunogen sequence: YAWIYYHMGR LSEAQAYVDK VRQVCQKFAN PYSMECPELE CEEGWTRLKC GRNERAKMCF EKALEEKPKD PECSSGMAIA MFRLEEKPEK QFSVDALKQA M.

IFN-induced antiviral protein which acts as an inhibitor of cellular as well as viral processes, cell migration, proliferation, signaling, and viral replication. Enhances MAVS-mediated host antiviral responses by serving as an adapter bridging TBK1 to MAVS which leads to the activation of TBK1 and phosphorylation of IRF3 and phosphorylated IRF3 translocates into nucleus to promote antiviral gene transcription. Exhibits an antiproliferative activity via the up-regulation of cell cycle negative regulators CDKN1A/p21 and CDKN1B/p27. Normally, CDKN1B/p27 turnover is regulated by COPS5, which binds CDKN1B/p27 in the nucleus and exports it to the cytoplasm for ubiquitin-dependent degradation. IFIT3 sequesters COPS5 in the cytoplasm, thereby increasing nuclear CDKN1B/p27 protein levels. Upregulates CDKN1A/p21 by downregulating MYC, a repressor of CDKN1A/p21. Can negatively regulate the apoptotic effects of IFIT2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFIT3
Gene Accession No. O14879, Q64345
Gene Alias cig41; CIG49; CIG-49; Garg49; GARG-49; Glucocorticoid-attenuated response gene 49 protein; Ifi49; IFI60; IFI-60K; IFIT3; IFIT-3; IFIT4; IFIT-4; interferon induced protein with tetratricopeptide repeats 3; interferon-induced 60 kDa protein; Interferon-induced protein with tetratricopeptide repeats 3; interferon-induced protein with tetratricopeptide repeats 4; interferon-stimulated protein 60; IRG2; Isg49; ISG60; ISG-60; P49; P60; retinoic acid-induced gene G protein; RIG-G; RP11-149I23.4
Gene Symbols IFIT3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of mouse IFIT3 (NP_0346311).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15959, 309526, 3437
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.