Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP158887 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP158887 100 μL
NBP15888720 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP158887 Supplier Novus Biologicals Supplier No. NBP158887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFIT5 Polyclonal specifically detects IFIT5 in Human samples. It is validated for Western Blot.

Specifications

Antigen IFIT5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q13325
Gene Alias FLJ53857, IFIT-5, interferon-induced protein with tetratricopeptide repeats 5, Retinoic acid- and interferon-inducible 58 kDa protein, retinoic acid- and interferon-inducible protein (58kD), RI58FLJ92678
Gene Symbols IFIT5
Host Species Rabbit
Immunogen Synthetic peptides corresponding to IFIT5(interferon-induced protein with tetratricopeptide repeats 5) The peptide sequence was selected from the middle region of IFIT5. Peptide sequence ITVYRLDDSDREGSVKSFSLGPLRKAVTLNPDNSYIKVFLALKLQDVHAE.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24138
Test Specificity Expected identity based on immunogen sequence: Human: 100%; Bovine: 92%; Canine: 92%; Pig: 92%; Equine: 78%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.