Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFIT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106800 Supplier Invitrogen™ Supplier No. PA5106800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFIT5.

Interferon-induced RNA-binding protein involved in the human innate immune response. Has a broad and adaptable RNA structure recognition important for RNA recognition specificity in antiviral defense. Binds precursor and processed tRNAs as well as poly-U-tailed tRNA fragments (PubMed:25092312, PubMed:23317505, PubMed:23774268). Specifically binds single-stranded RNA bearing a 5'-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs. Single-stranded PPP-RNAs, which lack 2'-O-methylation of the 5' cap and bear a 5'-triphosphate group instead, are specific from viruses, providing a molecular signature to distinguish between self and non-self mRNAs by the host during viral infection. Directly binds PPP-RNA in a non-sequence-specific manner (PubMed:23334420). Also recognizes and selectively binds AT-rich dsDNA (PubMed:23774268). Additionally, as a mediator in innate immunity, regulates positively IKK-NFKB signaling by sinergizing the recruitment of IKK to MAP3K7 (PubMed:26334375). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFIT5
Gene Accession No. Q13325
Gene Alias IFIT5; IFIT-5; interferon induced protein with tetratricopeptide repeats 5; interferon-induced 58 kDa protein; Interferon-induced protein with tetratricopeptide repeats 5; ISG58; P58; Retinoic acid- and interferon-inducible 58 kDa protein; retinoic acid- and interferon-inducible protein (58kD); RI58
Gene Symbols IFIT5
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFIT5(Accession Q13325), corresponding to amino acid residues E170-F220.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24138
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.