Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFITM1 Antibody (021), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289230AF594
Description
IFITM1 Monoclonal specifically detects IFITM1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
IFITM1 | |
Monoclonal | |
Alexa Fluor 594 | |
CD225 antigen, CD225Leu-13 antigen, IFI17LEU13,9-27, interferon induced transmembrane protein 1 (9-27), Interferon-induced protein 17, interferon-induced transmembrane protein 1, Interferon-inducible protein 9-27 | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human IFITM1. | |
0.1 mL | |
Cell Cycle and Replication | |
8519 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
21 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction