Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFITM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114745 Supplier Invitrogen™ Supplier No. PA5114745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFITM1.

IFITM1 (Interferon inducible transmembrane protein 1) is a member of the IFN-inducible transmembrane protein family. It is an essential mediator of interferon-gamma-induced antiproliferation and plays a role in the control of cell growth. IFITM1 is upregulated in several tumor types and may be useful as a tumor biomarker. Both mouse IFITM1 and IFITM3 are expressed on the cell surfaces of primordial germ cells in a developmentally-regulated manner. IFITM1 activity is required for primordial germ cell transit, and IFITM1 acts as a repulsive molecule by repelling non-IFITM1-expressing primordial germ cells from the mesoderm into the endoderm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFITM1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Ifitm1
Gene Accession No. P13164
Gene Alias 1110036C17Rik; 27-Sep; 9-27; CD225; dispanin subfamily A member 2a; DSPA2a; Fragilis protein 2; fragilis2; IFI17; IFITM1; ifitm-like protein 2; interferon induced transmembrane protein 1; interferon induced transmembrane protein 2 like; interferon-induced protein 17; Interferon-induced transmembrane protein 1; interferon-inducible protein 9-27; LEU13; Leu-13 antigen; Mil2; Mil-2; Mouse ifitm-like protein 2
Gene Symbols Ifitm1
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFITM1(Accession P13164), corresponding to amino acid residues M1-N43.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8519
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.