Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFITM1 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

$293.50 - $565.00

Specifications

Antigen IFITM1
Clone 001
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543443
View Documents
Invitrogen™
MA543443
50 μL
Each for $293.50
Only null left
Add to Cart
 
PIMA543108
GREENER_CHOICE
View Documents
Invitrogen™
MA543108
100 μL
Each for $565.00
Only null left
Add to Cart
 
Description

Description

Specificity: Human IFITM1

IFITM1 (Interferon inducible transmembrane protein 1) is a member of the IFN-inducible transmembrane protein family. It is an essential mediator of interferon-gamma-induced antiproliferation and plays a role in the control of cell growth. IFITM1 is upregulated in several tumor types and may be useful as a tumor biomarker. Both mouse IFITM1 and IFITM3 are expressed on the cell surfaces of primordial germ cells in a developmentally-regulated manner. IFITM1 activity is required for primordial germ cell transit, and IFITM1 acts as a repulsive molecule by repelling non-IFITM1-expressing primordial germ cells from the mesoderm into the endoderm.
Specifications

Specifications

IFITM1
1 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with no preservative
1110036C17Rik; 27-Sep; 9-27; CD225; dispanin subfamily A member 2a; DSPA2a; Fragilis protein 2; fragilis2; IFI17; IFITM1; ifitm-like protein 2; interferon induced transmembrane protein 1; interferon induced transmembrane protein 2 like; interferon-induced protein 17; Interferon-induced transmembrane protein 1; interferon-inducible protein 9-27; LEU13; Leu-13 antigen; Mil2; Mil-2; Mouse ifitm-like protein 2
Ifitm1
IgG
Protein A
Antibody
001
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
P13164
8519
A synthetic peptide corresponding to the center region of the Human IFITM1.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Ifitm1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.