Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFITM2/3 Mouse anti-Human, Clone: 2E8D12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | IFITM2/3 |
---|---|
Clone | 2E8D12 |
Concentration | 0.85 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6518
![]() ![]() |
Proteintech Group Inc
660811IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes both IFITM2 and IFITM3.
IFN-induced antiviral protein that mediates cellular innate immunity to at least three major human pathogens, namely influenza A H1N1 virus, West Nile virus (WNV), and dengue virus (WNV), by inhibiting the early step(s) of replication.Specifications
IFITM2/3 | |
0.85 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
1 8U, IFITM3, IP15 | |
IFITM2, IFITM3 | |
IgG1 | |
Protein G | |
Antibody |
2E8D12 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q01628, Q01629 | |
10410, 10581 | |
IFITM2/3 Fusion Protein Ag17863 | |
Primary | |
-20°C | |
IFITM3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title