Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFITM2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299453MFV450
Description
IFITM2 Polyclonal antibody specifically detects IFITM2 in Human samples. It is validated for Western BlotSpecifications
IFITM2 | |
Polyclonal | |
Western Blot | |
1-8D, interferon induced transmembrane protein 2 (1-8D), interferon-induced transmembrane protein 2, Interferon-inducible protein 1-8D | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IFITM2. | |
0.1 mL | |
Cancer, Immunology, Innate Immunity | |
10581 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction