Learn More
Invitrogen™ IFITM2 Polyclonal Antibody

Description
IFITM2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | IFITM2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | IFITM2 |
| Gene Accession No. | Q01629 |
| Gene Alias | 1-8D; dispanin subfamily A member 2c; DSPA2c; IFITM2; ifm2; interferon induced transmembrane protein 2; interferon-induced transmembrane protein 2; Interferon-inducible protein 1-8D |
| Gene Symbols | IFITM2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.