Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFLTD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282095MFV450
Description
IFLTD1 Polyclonal antibody specifically detects IFLTD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| IFLTD1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ36004, intermediate filament tail domain containing 1, intermediate filament tail domain-containing protein 1, Pas1c1 | |
| IFLTD1 antibody was raised against an 19 amino acid synthetic peptide near the center of human IFLTD1. The immunogen is located within amino acids 200 - 250 of IFTLD1. Amino Acid Squence: EAKHQPPSDFLWKEQDKFR | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 160492 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction