Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFLTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543952 Supplier Invitrogen™ Supplier No. PA543952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PPTVFPNRSP WCQNPYVSAH PYCPLIEPHN TSTAGGRLDR QPRSRSTRPN Sequence homology: Human: 100%.

IFLTD1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytoplasm; intermediate filament; nuclear envelope; structural molecule activity; cell population proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFLTD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Lmntd1
Gene Accession No. Q8N9Z9
Gene Alias 4933403M22Rik; AI482300; IFLTD1; intermediate filament tail domain containing 1; intermediate filament tail domain-containing protein 1; lamin A-related sequence 1; lamin tail domain containing 1; lamin tail domain-containing protein 1; Lamin-A-related sequence 1 protein; Lmna1; Lmna-rs1; Lmntd1; PAS1C1; RGD1560077
Gene Symbols Lmntd1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human IFLTD1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160492
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.