Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFLTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561123 Supplier Invitrogen™ Supplier No. PA561123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEAKHQPPSD FLWKEQDKFR ASPDCITILC KPNGQAIAWY TPIHWKQAWE KLDADVEFNR CSVVSPTFRK RVFQWTASTA TITKE Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 65%.

IFLTD1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytoplasm; intermediate filament; nuclear envelope; structural molecule activity; cell population proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFLTD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lmntd1
Gene Accession No. Q8N9Z9
Gene Alias 4933403M22Rik; AI482300; IFLTD1; intermediate filament tail domain containing 1; intermediate filament tail domain-containing protein 1; lamin A-related sequence 1; lamin tail domain containing 1; lamin tail domain-containing protein 1; Lamin-A-related sequence 1 protein; Lmna1; Lmna-rs1; Lmntd1; PAS1C1; RGD1560077
Gene Symbols Lmntd1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFLTD1. Recombinant protein control fragment (Product #RP-97781).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160492
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.