Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha 1 Antibody (AB5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073047
Description
IFN-alpha 1 Monoclonal specifically detects IFN-alpha 1 in Human samples. It is validated for Western Blot, Sandwich ELISA.Specifications
IFN-alpha D | |
Monoclonal | |
Unconjugated | |
IFL, IFN, IFNA13, IFNAalphaIFN-alpha-1/13, IFN-ALPHA, IFN-alpha 1b, IFN-alphaD, interferon alpha 1b, interferon alpha-1/13, Interferon alpha-D, interferon, alpha 1, LeIF D, MGC138207, MGC138505, MGC138507 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Interferon Alpha (AB5) | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, ELISA | |
AB5 | |
Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
IFNA1 | |
recombinant human interferon alpha-A. | |
0.2 mg | |
Cancer, Immunology, Innate Immunity | |
3439 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction