Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha B2/IFN-alpha 8 Antibody (003), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289440DL594
Description
IFN-alpha B2/IFN-alpha 8 Monoclonal antibody specifically detects IFN-alpha B2/IFN-alpha 8 in Human samples. It is validated for ELISASpecifications
IFN-alpha B2/IFN-alpha 8 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
003 | |
ELISA | |
IFN-alpha-8, IFN-alphaB, interferon alpha type 201, interferon alpha-8, interferon alpha-B', Interferon alpha-B, Interferon alpha-B2, interferon, alpha 8, LeIF B | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-alpha B2/IFN-alpha 8 (NP_002161.2; Cys24-Glu189). | |
0.1 mL | |
Cancer, Immunology, Innate Immunity | |
3445 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction