Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha/beta R1 Antibody (85228), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB245PECY7
Description
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Flow CytometrySpecifications
IFN-alpha/beta R1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
85228 | |
Flow Cytometry | |
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IFN-alpha/beta R1, Lys28-Lys436, Accession # AAA52730 | |
0.1 mL | |
Neuroscience, Phospho Specific | |
3454 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction