Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha/beta R1 Antibody (MM0336-8H64), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211731AF647
Description
IFN-alpha/beta R1 Monoclonal specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
IFN-alpha/beta R1 | |
Monoclonal | |
Alexa Fluor 647 | |
AAA52730 | |
IFNAR1 | |
Human recombinant IFN-alpha/beta R1 (extracellular domain) | |
0.1 mL | |
Neuroscience, Phospho Specific | |
3454 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
MM0336-8H64 | |
Western Blot, Flow Cytometry | |
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction