Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IFN-alpha/beta R1 Antibody Pair [HRP]
SDP

Catalog No. NB009863 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009863 15 Plates
NB009864 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB009863 Supplier Novus Biologicals™ Supplier No. NBP27941215PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IFN-alpha/beta R1.

Specifications

Assay Range 78 pg/mL
Assay Sensitivity 78.13-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IFN-alpha/beta R1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Gene ID (Entrez) 3454
Gene Symbol IFNAR1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 95 ng of recombinant IFN-alpha/beta R1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-IFN-alpha/beta R1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL mouse anti-IFN-alpha/beta R1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-protector, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Neuroscience, Phospho Specific
    Storage Requirements Storage is content dependent.
    Target IFN-alpha/beta R1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.