Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha/beta R1 Mouse anti-Human, Clone: 8H46, Novus Biologicals™

Mouse Monoclonal Antibody
$244.00 - $646.00
Specifications
Antigen | IFN-alpha/beta R1 |
---|---|
Clone | 8H46 |
Concentration | LYOPH |
Applications | Flow Cytometry, Western Blot, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019665
![]() |
Novus Biologicals
NBP2362580.02MG |
0.02 mg |
Each for $244.00
|
|
|||||
NBP23625805
![]() |
Novus Biologicals
NBP2362580.05MG |
0.05 mg |
Each for $401.00
|
|
|||||
NBP236258
![]() |
Novus Biologicals
NBP236258 |
0.1 mg |
Each for $646.00
|
|
|||||
Description
IFN-alpha/beta R1 Monoclonal specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
IFN-alpha/beta R1 | |
LYOPH | |
Monoclonal | |
Mouse | |
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
IFNAR1 | |
IgG2 |
8H46 | |
Flow Cytometry, Western Blot, CyTOF | |
Unconjugated | |
Human | |
3454 | |
recombinant human INF alpha/beta R1 EC domain | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title