Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha/beta R2 Antibody (122), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289447MFV450
Description
IFN-alpha/beta R2 Monoclonal antibody specifically detects IFN-alpha/beta R2 in Human samples. It is validated for Flow CytometrySpecifications
IFN-alpha/beta R2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
122 | |
Flow Cytometry | |
beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFNARB, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-alpha/beta R2 (NP_997468.1; Met1-Lys243). | |
0.1 mL | |
Cancer | |
3455 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction