Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha C/IFN-alpha 10 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | IFN-alpha C/IFN-alpha 10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
IFN-alpha C/IFN-alpha 10 Polyclonal antibody specifically detects IFN-alpha C/IFN-alpha 10 in Rat samples. It is validated for Western BlotSpecifications
IFN-alpha C/IFN-alpha 10 | |
Polyclonal | |
Purified | |
RUO | |
Rat | |
IFN-alpha-10, IFN-alphaC, interferon alpha-10, Interferon alpha-6L, Interferon alpha-C, interferon, alpha 10, LeIF C, MGC119878, MGC119879 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 24-100 of human IFN-alpha C/IFN-alpha 10 (NP_002162.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Immunology, Innate Immunity | |
PBS, 50% glycerol, pH7.3 | |
3446 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title