Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha G/IFNA5 Antibody (126), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289437AF750
Description
IFN-alpha G/IFNA5 Monoclonal antibody specifically detects IFN-alpha G/IFNA5 in Human samples. It is validated for ELISASpecifications
IFN-alpha G/IFNA5 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3442 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
126 | |
ELISA | |
IFN-alpha-5, IFN-alphaG, INFA5, interferon alpha-5, Interferon alpha-61, Interferon alpha-G, interferon, alpha 5, LeIF G | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-alpha G/IFNA5 (P01569; Met1-Glu189). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction