Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha H2/IFNA14 Antibody (065), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290671IR
Description
IFN-alpha H2/IFNA14 Monoclonal specifically detects IFN-alpha H2/IFNA14 in Mouse samples. It is validated for ELISA.Specifications
IFN-alpha H2/IFNA14 | |
Monoclonal | |
DyLight 755 | |
IFN-alpha-14, IFN-alphaH, interferon alpha-14, Interferon alpha-H, Interferon lambda-2-H, interferon, alpha 14, LeIF H, LEIF2H, MGC125756, MGC125757 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFN-alpha H2/IFNA14 (NP_996858.1; Cys24-Lys189). | |
0.1 mL | |
Cytokine Research | |
3448 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
65 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction