Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha I/IFNA17 Antibody (995819) [Biotin], Novus Biologicals™
SDP

Catalog No. FAB100801B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB100801B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB100801B Supplier R&D Systems Supplier No. FAB100801B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IFN-alpha I/IFNA17 Monoclonal specifically detects IFN-alpha I/IFNA17 in Human samples. It is validated for Neutralization.

Specifications

Antigen IFN-alpha I/IFNA17
Applications Neutralization
Classification Monoclonal
Clone 995819
Conjugate Biotin
Formulation PBS
Gene Alias IFNA, IFN-alpha-17, IFN-alphaI, INFA, interferon alpha-17, Interferon alpha-88, Interferon alpha-I', Interferon alpha-T, interferon alpha-WA, interferon, alpha 17, LeIF I, LEIF2C1
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human IFN-alpha I, Met1-Asp189, Accession # P01571
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3451
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.