Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131156
Description
IFN-alpha K Polyclonal specifically detects IFN-alpha K in Human samples. It is validated for Western Blot.Specifications
IFN-alpha K | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
IFNA6 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human IFN-alpha K. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
3443 | |
Human | |
IgG |
Western Blot | |
1.0mg/mL | |
Western Blot 1:100-1:1000 | |
IFNA6, IFN-alpha 6, IFNalpha K, IFN-alpha K, IFN-kappa, interferon kappa, interferon, kappa, RP11-27J8.1, UNQ6124/PRO20084 | |
Rabbit | |
22 kDa | |
100 μL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction