Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha K Antibody (OTI2A10), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271007AF700
Description
IFN-alpha K Monoclonal specifically detects IFN-alpha K in Human samples. It is validated for Western Blot.Specifications
IFN-alpha K | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IFNA6 | |
Full length recombinant protein of human IFN-alpha K (NP_066282) produced in E.coli. | |
0.1 mL | |
3443.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2A10 | |
Western Blot | |
IFNA6, IFN-alpha 6, IFNalpha K, IFN-alpha K, IFN-kappa, interferon kappa, interferon, kappa, RP11-27J8.1, UNQ6124/PRO20084 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction