Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-beta Antibody (040), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289612C
Description
IFN-beta Monoclonal antibody specifically detects IFN-beta in Human samples. It is validated for ELISASpecifications
IFN-beta | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
040 | |
ELISA | |
Fibroblast interferon, IFBIFNB, IFF, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-beta (NP_002167.1; Met1-Asn187). | |
0.1 mL | |
Apoptosis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3456 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction