Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFN beta Monoclonal Antibody (MMHB-3)

Catalog No. PI214001
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI214001 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI214001 Supplier Invitrogen™ Supplier No. 214001
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: Neutralizes human interferon beta. For best results in experiments involving neutralization of the anti-viral effect of human interferon beta we recommend using sheep polyclonal antibody to human interferon beta (Product # 314001). Activity: Interferon was titrated with the use of the cytopathic effect inhibition assay, where 1 unit/mL of interferon was necessary to produce an endpoint of 50%. One neutralization unit is defined as the amount of serum required to neutralize one unit of human IFN beta to an endpoint of 50%. Purity is >95%. Endotoxin level <1 EU/ug. Store at -20°C for short-term use (less than 6 months). For long-term storage, aliquot and store at -70°C or below for retention of full activity, avoiding freeze/thaw cycles. Packaged by and distributed for PBL Assay Science.

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN beta
Applications ELISA, Neutralization
Classification Monoclonal
Clone MMHB-3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene Ifnb1
Gene Accession No. P01574
Gene Alias Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1
Gene Symbols Ifnb1
Host Species Mouse
Immunogen human interferon beta.
Purification Method Size-exclusion chromatography, Ion-exchange chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.