Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFN beta Monoclonal Antibody (A1 (IFNb)), Functional Grade, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7069953
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-148-12 50 μg
50-148-13 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-148-12 Supplier Invitrogen™ Supplier No. 16997881
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The A1 monoclonal antibody reacts with human IFN-beta. The A1 antibody is a neutralizing antibody. IFN-beta is classified as a type I IFN which is a group of structurally and functionally related proteins demonstrating anti-viral, anti-parasitic, and anti-proliferative activities. Applications Reported: The A1 antibody has been reported for use in cytokine neutralization. Applications Tested: The Functional Grade Purified A1 antibody has been tested in bioassay for neutralization of IFN-beta bioactivity. The A1 antibody at 3-6 μg/mL has been found to inhibit by 50% the biological effects of 50 pg/mL human IFN-beta (ND50), in an EMCV assay of A549 cell protection. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the Best Protocols™ section. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN beta
Applications Functional Assay, Neutralization
Classification Monoclonal
Clone A1 (IFNb)
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Ifnb1
Gene Accession No. P01574
Gene Alias Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1
Gene Symbols Ifnb1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.