Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFN epsilon Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546164
Description
Peptide sequence: IFSLFRANIS LDGWEENHTE KFLIQLHQQL EYLEALMGLE AEKLSGTLGS Sequence homology: Cow: 93%; Human: 100%; Pig: 79% PA5-46164 was successfully used to detect IFNE in endometrial tissue sections by IHC.
Type I interferon required for maintaining basal levels of IFN-regulated genes, including 2'-5'-oligoadenylate synthetase, IRF7 and ISG15, in the female reproductive tract. Directly mediates protection against viral and bacterial genital infections. [UniProt].
Specifications
IFN epsilon | |
Polyclonal | |
Unconjugated | |
IFNE | |
IFNE; IFN-E; Ifne1; IFN-epsilon; Ifnt1; Ifn-tau-1; Infe1; Interferon; Interferon epsilon; interferon epsilon 1; interferon epsilon-1; Interferon tau-1; interferon, epsilon; interferon-epsilon; PRO655; UNQ360/PRO655 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
338376 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WN2 | |
IFNE | |
Synthetic peptide directed towards the middle region of human IFNE. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction