Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma Antibody (121), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289964AF647
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for ELISA.Specifications
IFN-gamma | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
121 | |
ELISA | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-gamma (NP_000610.2; Met 1-Gln 166). | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
3458 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction