Learn More
IFN-gamma Antibody (345025), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2300U
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
IFN-gamma | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Bovine | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
345025 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
E. coli-derived recombinant bovine IFN-gamma , Gln24-Thr166, Accession # NP_776511 | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
3458 | |
Store at 4C in the dark. | |
IgG2a |