Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma Antibody (37895R), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB485RK
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Mouse samples. It is validated for Neutralization, Flow Cytometry (Intracellular Staining)Specifications
IFN-gamma | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Neutralization, Flow Cytometry (Intracellular Staining) | |
37895R | |
Neutralization, Intracellular Staining by Flow Cytometry | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
E. coli-derived recombinant mouse IFN-gamma | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
3458 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction