Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma Antibody (D9D10), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281044AF405
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
IFN-gamma | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
IFNG | |
FPLC-purified natural human IFN-gamma. Refer to the original article [PMID: 3114127] | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
3458 | |
Human, Baboon | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
D9D10 | |
Western Blot, Flow Cytometry, ELISA | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Binds human IFN-gamma, a dimerized soluble cytokine that is the only member of the type II class of interferons. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction