Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma Antibody (N3-P3A5*A10), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250435IR
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
IFN-gamma | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IFNG | |
Ovalbumin-conjugated synthetic peptide | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
3458 | |
Store at 4C in the dark. | |
IgG2a λ |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
N3-P3A5*A10 | |
Western Blot, ELISA, Immunohistochemistry | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction