Learn More
Invitrogen™ IFN gamma Monoclonal Antibody (MD-1), eBioscience™

Description
Description: The MD-1 antibody reacts with human, rhesus, cynomolgus and baboon interferon-gamma (IFN gamma), the signature Th1 cytokine, which is produced mainly by activated T cells, NK cells and B cells. IFN gamma is a dimeric protein. IFN gamma has antiviral and antiparasitic activities and also inhibits the proliferation of normal and transformed cells. Applications Reported: The MD-1 antibody has been reported for use in intracellular flow cytometric analysis, cytokine neutralization, immunohistochemical staining, ELISA, and ELISPOT. (Fluorochrome conjugated MD-1 is recommended for use in intracellular flow cytometry.) (Please use Functional Grade purified MD-1, Product # 16-7317, in functional assays). Applications Tested: The MD-1 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human interferon gamma (IFN gamma) in combination with the biotinylated 4S.B3 (Product # 13-7319-81) antibody for detection and recombinant human IFN gamma (39-8319) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. The FG Purified format of the MD-1 antibody has been tested as the capture antibody in an ELISPOT assay (in conjunction with the biotinylated 4S.B3 antibody) and for neutralization of IFN gamma biological activity. A suitable range of c...
Specifications
Specifications
| Antigen | IFN gamma |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunostaining, Western Blot |
| Classification | Monoclonal |
| Clone | MD-1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | IFNG |
| Gene Accession No. | P01579 |
| Gene Alias | BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.